Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromopyridine N-oxide, 97%

$216.97 - $667.50

Chemical Identifiers

Chemical Name or Material 3-Bromopyridine N-oxide
CAS 2402-97-3
Molecular Formula C5H4BrNO
Formula Weight 174
MDL Number MFCD01646056
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2594006

  SDS
alfa aesar™
H2594006
5g Each for $216.97

AAH2594014

  SDS
alfa aesar™
H2594014
25g Each for $667.50
Description & Specifications

Specifications

Formula Weight 174
Molecular Formula C5H4BrNO
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Boiling Point 130°C (2mmHg)
CAS 2402-97-3
MDL Number MFCD01646056
Quantity 5g
Melting Point 30° to 40°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material 3-Bromopyridine N-oxide