missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromopyridine 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107411000

 View more versions of this product

Catalog No. AC107411000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Bromopyridine
626-55-1
Authentic
Glass bottle
100mL
≥98.5%
99%
(C5H4N)Br
MFCD00006373
99%
98.5
100.0
1.5690 to 1.5710 (20°C, 589nm)
Liquid
Clear colorless to yellow or slightly brown
C5H4BrN
158
20, 233

Chemical Identifiers

626-55-1
100.0
C5H4BrN
158
3-Bromopyridine, 99%
98.5
99%
(C5H4N)Br
MFCD00006373
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-952-0

TSCA : TSCA