Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromopyridine-4-carboxylic acid, 97%

$104.99 - $290.92

Chemical Identifiers

Chemical Name or Material 3-Bromopyridine-4-carboxylic acid
CAS 13959-02-9
Molecular Formula C6H4BrNO2
Formula Weight 202.01
MDL Number MFCD00040944
Synonym 3-Bromoisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3104703

  SDS
alfa aesar™
H3104703
1g Each for $290.92

AAH31047MD

  SDS
alfa aesar™
H31047MD
250mg Each for $104.99
Description & Specifications

Specifications

Formula Weight 202.01
Chemical Name or Material 3-Bromopyridine-4-carboxylic acid
Assay 97%
Molecular Formula C6H4BrNO2
MDL Number MFCD00040944
CAS 13959-02-9
Melting Point ≈235°C
Synonym 3-Bromoisonicotinic acid
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.