missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromopropylamine Hydrobromide 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 107380250

 View more versions of this product

Catalog No. AC107380250


 

Specifications Description & Specifications

Specifications

≥97.5%
218.93
White to off-white
04, 149
Glass bottle
C3H8BrN·HBr
MFCD00012912
3-Bromopropylamine hydrobromide
98%
Powder or Chunks
25g
171° to 175°C
100.0
Br(CH2)3NH2·HBr
98%
97.5
5003-71-4
Authentic

Chemical Identifiers

5003-71-4
100.0
C3H8BrN·HBr
218.93
3-Bromopropylamine hydrobromide, 98%
97.5
98%
Br(CH2)3NH2·HBr
MFCD00012912
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-675-0

RTECSNumber : UH9925000

TSCA : TSCA