Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Bromopropyl)triphenylphosphonium bromide, 98%

$61.26 - $172.41

Chemical Identifiers

Chemical Name or Material (3-Bromopropyl)triphenylphosphonium bromide
CAS 3607-17-8
Molecular Formula BrCH2CH2CH2P(C6H5)3Br
Formula Weight 464.19
MDL Number MFCD00011866
Beilstein 6422974
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1304114

  SDS
alfa aesar™
A1304114
25g Each for $61.26

AAA1304122

  SDS
alfa aesar™
A1304122
100g Each for $172.41
Description & Specifications

Specifications

Formula Weight 464.19
Chemical Name or Material (3-Bromopropyl)triphenylphosphonium bromide
Assay 98%
Molecular Formula BrCH2CH2CH2P(C6H5)3Br
MDL Number MFCD00011866
CAS 3607-17-8
Melting Point 225° to 229°C
Beilstein 6422974
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.