missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromopropionyl Chloride, TCI America™

$90.14 - $586.67

Chemical Identifiers

CAS 15486-96-1
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.418
MDL Number MFCD00000746
InChI Key IHBVNSPHKMCPST-UHFFFAOYSA-N
Synonym 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
PubChem CID 84944
IUPAC Name 3-bromopropanoyl chloride
SMILES C(CBr)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B206525G View Documents TCI America
B206525G
25g Each for $90.14
 
B2065250G View Documents TCI America
B2065250G
250g Each for $586.67
 
Specifications

Chemical Identifiers

15486-96-1
171.418
IHBVNSPHKMCPST-UHFFFAOYSA-N
84944
C(CBr)C(=O)Cl
C3H4BrClO
MFCD00000746
3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
3-bromopropanoyl chloride

Specifications

Undesignated
15486-96-1
MFCD00000746
3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
C(CBr)C(=O)Cl
171.418
171.42
≥95.0% (GC,T)
25g
C3H4BrClO
3265
IHBVNSPHKMCPST-UHFFFAOYSA-N
3-bromopropanoyl chloride
84944
Clear Liquid at 20°C
3-Bromopropionyl Chloride
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator