Please login to your online account to display your discounted pricing

3-bromopropionaldehyde dimethylacetal, 95%, ACROS Organics™

$67.25 - $67.25

Chemical Identifiers

Chemical Name or Material 3-Bromopropionaldehyde dimethylacetal
CAS 36255-44-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H11BrO2
Linear Formula Br(CH2)2CH(OCH3)2
Formula Weight 183.04
MDL Number MFCD00013245
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376260010

  SDS
thermo fisher scientific
AC376260010
Glass bottle 1g Each for $67.25

AC376260050

  SDS
thermo fisher scientific
AC376260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 183.04
Quantity 1g
Color Clear colorless to pale yellow
Density 1.341g/mL
CAS Min % 94.0
CAS Max % 100.0
Linear Formula Br(CH2)2CH(OCH3)2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C5H11BrO2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Packaging Glass bottle
Refractive Index 1.4460 to 1.4500 (20°C, 589nm)
Synonym 1-Bromo-3, 3-dimethoxypropane
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 3-Bromopropionaldehyde dimethylacetal
MDL Number MFCD00013245
CAS 36255-44-4
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.