Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromopropionaldehyde dimethyl acetal, stab., tech. 90%

$118.22 - $118.22

Chemical Identifiers

Chemical Name or Material 3-Bromopropionaldehyde dimethyl acetal, stab.
CAS 36255-44-4
Purity Grade Technical Grade
Molecular Formula C5H11BrO2
Formula Weight 183.04
MDL Number MFCD00013245
UN Number UN1993
Beilstein 1697640
Synonym 1-Bromo-3,3-dimethoxypropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2015006

  SDS
alfa aesar™
L2015006
5g Each for $118.22

AAL2015014

  SDS
alfa aesar™
L2015014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.04
Molecular Formula C5H11BrO2
Beilstein 1697640
Synonym 1-Bromo-3,3-dimethoxypropane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 45°C (113°F)
Chemical Name or Material 3-Bromopropionaldehyde dimethyl acetal, stab.
Purity Grade Technical Grade
Assay 90%
MDL Number MFCD00013245
CAS 36255-44-4
UN Number UN1993
Boiling Point 58° to 60°C (17mmHg)
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Applications
3-Bromopropionaldehyde dimethyl acetal was used in the synthesis of spiro cyclobutanones.

Solubility
Not miscible or difficult to mix with water.

Notes
Store under refrigerated condition. Store away from oxidizing agents and protect from heat.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C