missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromophenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2351014

 View more versions of this product

Catalog No. AAB2351014



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromophenylhydrazine hydrochloride
∼225°C (decomposition)
MFCD00012933
3565829
RPYIPFXHIKXRKS-UHFFFAOYSA-N
(3-bromophenyl)hydrazine;hydrochloride
33757
98%
27246-81-7
C6H8BrClN2
25 g
3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
[H+].[Cl-].NNC1=CC=CC(Br)=C1
223.50
223.51

Chemical Identifiers

27246-81-7
223.50
RPYIPFXHIKXRKS-UHFFFAOYSA-N
33757
[H+].[Cl-].NNC1=CC=CC(Br)=C1
C6H8BrClN2
MFCD00012933
3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
(3-bromophenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 248-362-0

RTECSNumber : MV0815000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only