Learn More
3-Bromophenylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 358940010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Bromophenylboronic acid | |
96.0 | |
Authentic | |
Glass bottle | |
BrC6H4B(OH)2 | |
1g | |
Solubility in water: 25g/L | |
B(C1=CC(=CC=C1)Br)(O)O | |
200.83 | |
200.83 |
89598-96-9 | |
100.0 | |
97% | |
C6H6BBrO2 | |
MFCD00239386 | |
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid | |
AFSSVCNPDKKSRR-UHFFFAOYSA-N | |
(3-bromophenyl)boronic acid | |
2734318 | |
97% |
Chemical Identifiers
89598-96-9 | |
200.83 | |
AFSSVCNPDKKSRR-UHFFFAOYSA-N | |
2734318 | |
B(C1=CC(=CC=C1)Br)(O)O |
C6H6BBrO2 | |
MFCD00239386 | |
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid | |
(3-bromophenyl)boronic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
RUO â Research Use Only