Please login to your online account to display your discounted pricing

3-Bromophenylacetone, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC350600250

 View more versions of this product

Catalog No. AC350600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 213.08
Physical Form Liquid
Quantity 25g
Color Clear light yellow to dark yellow
CAS Max % 100.0
CAS Min % 98.0
Chemical Name or Material 3-Bromophenylacetone
Molecular Formula C9H9BrO
CAS 21906-32-1
Assay Percent Range 98+%
MDL Number MFCD00210400
Refractive Index 1.5570 to 1.5600 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Bromophenylacetone
CAS 21906-32-1
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H9BrO
Formula Weight 213.08
MDL Number MFCD00210400