missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromophenylacetone, 98+%, ACROS Organics™

Chemical Identifiers

CAS 21906-32-1
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00210400
InChI Key FQIZFAJMBXZVOL-UHFFFAOYSA-N
Synonym 1-3-bromophenyl propan-2-one, 3-bromophenylacetone, 1-3-bromophenyl-2-propanone, 1-3-bromophenyl acetone, 2-propanone, 1-3-bromophenyl, 3-bromophenyl acetone, ksc492m0h, 1-3-bromo-phenyl propan-2-one
PubChem CID 2734093
IUPAC Name 1-(3-bromophenyl)propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC350600250 SDS View Documents Acros Organics
AC350600250
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350600050 SDS View Documents Acros Organics
350600050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

21906-32-1
213.074
FQIZFAJMBXZVOL-UHFFFAOYSA-N
2734093
CC(=O)CC1=CC(=CC=C1)Br
C9H9BrO
MFCD00210400
1-3-bromophenyl propan-2-one, 3-bromophenylacetone, 1-3-bromophenyl-2-propanone, 1-3-bromophenyl acetone, 2-propanone, 1-3-bromophenyl, 3-bromophenyl acetone, ksc492m0h, 1-3-bromo-phenyl propan-2-one
1-(3-bromophenyl)propan-2-one

Specifications

Authentic
1.5570 to 1.5600 (20°C, 589nm)
25g
98.0
98+%
MFCD00210400
FQIZFAJMBXZVOL-UHFFFAOYSA-N
1-(3-bromophenyl)propan-2-one
2734093
Liquid
3-Bromophenylacetone
Glass bottle
Yellow
21906-32-1
100.0
C9H9BrO
1-3-bromophenyl propan-2-one, 3-bromophenylacetone, 1-3-bromophenyl-2-propanone, 1-3-bromophenyl acetone, 2-propanone, 1-3-bromophenyl, 3-bromophenyl acetone, ksc492m0h, 1-3-bromo-phenyl propan-2-one
CC(=O)CC1=CC(=CC=C1)Br
213.074
213.08
≥98.0%