Please login to your online account to display your discounted pricing

3-Bromophenylacetic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromophenylacetic acid
CAS 1878-67-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7BrO2
Linear Formula BrC6H4CH2COOH
Formula Weight 215.05
MDL Number MFCD00004330
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC296640025

  SDS
fisher scientific acros
296640025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder or Needles
Formula Weight 215.05
Quantity 2.5g
Color White
Melting Point 99° to 102°C
CAS Max % 100.0
CAS 1878-67-7
Assay Percent Range 98%
Infrared Spectrum Authentic
Chemical Name or Material 3-Bromophenylacetic acid
MDL Number MFCD00004330
CAS Min % 97.5
Linear Formula BrC6H4CH2COOH
Packaging Glass bottle
Molecular Formula C8H7BrO2