Please login to your online account to display your discounted pricing

3-Bromophenylacetic acid, 98%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  296640025

 View more versions of this product

Catalog No. AC296640025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Needles
Formula Weight 215.05
Quantity 2.5g
Color White
Melting Point 99° to 102°C
CAS Max % 100.0
CAS 1878-67-7
Assay Percent Range 98%
Infrared Spectrum Authentic
Chemical Name or Material 3-Bromophenylacetic acid
MDL Number MFCD00004330
CAS Min % 97.5
Linear Formula BrC6H4CH2COOH
Packaging Glass bottle
Molecular Formula C8H7BrO2

Specifications

Chemical Name or Material 3-Bromophenylacetic acid
CAS 1878-67-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7BrO2
Linear Formula BrC6H4CH2COOH
Formula Weight 215.05
MDL Number MFCD00004330