missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Bromophenyl)triphenylsilane, TCI America™

$128.34 - $381.95

Chemical Identifiers

CAS 185626-73-7
Molecular Formula C24H19BrSi
Molecular Weight (g/mol) 415.405
MDL Number MFCD28098175
InChI Key WTYKIOBQNRWDQP-UHFFFAOYSA-N
Synonym 3-Bromotetraphenylsilane
PubChem CID 85709247
IUPAC Name (3-bromophenyl)-triphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC(=CC=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B52221G View Documents TCI America
B52221G
1g Each for $381.95
 
B5222200MG View Documents TCI America
B5222200MG
200mg Each for $128.34
 
Specifications

Chemical Identifiers

185626-73-7
415.405
WTYKIOBQNRWDQP-UHFFFAOYSA-N
85709247
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC(=CC=C4)Br
C24H19BrSi
MFCD28098175
3-Bromotetraphenylsilane
(3-bromophenyl)-triphenylsilane

Specifications

White-Yellow
1g
C24H19BrSi
3-Bromotetraphenylsilane
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC(=CC=C4)Br
415.405
415.41
≥98.0% (GC)
137°C
185626-73-7
MFCD28098175
WTYKIOBQNRWDQP-UHFFFAOYSA-N
(3-bromophenyl)-triphenylsilane
85709247
Crystal-Powder at 20°C
(3-Bromophenyl)triphenylsilane
Safety and Handling

Safety and Handling

TSCA : No