missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L0987114

 View more versions of this product

Catalog No. AAL0987114



Specifications

Specifications

3-Bromophenyl isothiocyanate
Pungent
25g
C7H4BrNS
UN2922
3-bromophenyl isothiocyanate, m-bromophenyl isothiocyanate, benzene, 1-bromo-3-isothiocyanato, isothiocyanic acid, m-bromophenyl ester, 3-bromophenylisothiocyanate, 1-bromo-3-isothiocyanato-benzene, acmc-1chhg, m-bromphenylisothiocyanat, zmgmghnoacsmqn-uhfffaoysa, #
C1=CC(=CC(=C1)Br)N=C=S
214.08
214.09
>110°C (230°F)
255°C to 256°C
2131-59-1
MFCD00004803
2081173
ZMGMGHNOACSMQN-UHFFFAOYSA-N
1-bromo-3-isothiocyanatobenzene
16481
97%

Chemical Identifiers

2131-59-1
214.08
ZMGMGHNOACSMQN-UHFFFAOYSA-N
16481
C1=CC(=CC(=C1)Br)N=C=S
C7H4BrNS
MFCD00004803
3-bromophenyl isothiocyanate, m-bromophenyl isothiocyanate, benzene, 1-bromo-3-isothiocyanato, isothiocyanic acid, m-bromophenyl ester, 3-bromophenylisothiocyanate, 1-bromo-3-isothiocyanato-benzene, acmc-1chhg, m-bromphenylisothiocyanat, zmgmghnoacsmqn-uhfffaoysa, #
1-bromo-3-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : NX8350000

Documents