Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromophenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromophenyl isocyanate
CAS 23138-55-8
Molecular Formula C7H4BrNO
Formula Weight 198.03
MDL Number MFCD00002014
UN Number UN2206
Beilstein 2802537
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1229203

  SDS
alfa aesar™
L1229203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1229206

  SDS
alfa aesar™
L1229206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.03
Chemical Name or Material 3-Bromophenyl isocyanate
Assay 98%
Molecular Formula C7H4BrNO
MDL Number MFCD00002014
CAS 23138-55-8
UN Number UN2206
Beilstein 2802537
Boiling Point 220°C
Odor Pungent
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Quantity 1g
Flash Point 108°C (226°F)
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C