missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromophenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1229203

 View more versions of this product

Catalog No. AAL1229203



Specifications

Specifications

3-Bromophenyl isocyanate
Pungent
1g
C7H4BrNO
UN2206
3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
C1=CC(=CC(=C1)Br)N=C=O
198.019
198.03
108°C (226°F)
220°C
23138-55-8
MFCD00002014
2802537
VQVBCZQTXSHJGF-UHFFFAOYSA-N
1-bromo-3-isocyanatobenzene
141001
98%

Chemical Identifiers

23138-55-8
198.019
VQVBCZQTXSHJGF-UHFFFAOYSA-N
141001
C1=CC(=CC(=C1)Br)N=C=O
C7H4BrNO
MFCD00002014
3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
1-bromo-3-isocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents