Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromopentane, 97%

$337.51 - $337.51

Chemical Identifiers

Chemical Name or Material 3-Bromopentane
CAS 1809-10-5
Molecular Formula C5H11Br
Formula Weight 151.05
MDL Number MFCD00000158
UN Number UN1993
Beilstein 1730967
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1357606

  SDS
alfa aesar™
L1357606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1357614

  SDS
alfa aesar™
L1357614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1357622

  SDS
alfa aesar™
L1357622
100g Each for $337.51
Description & Specifications

Specifications

Formula Weight 151.05
Chemical Name or Material 3-Bromopentane
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1809-10-5
MDL Number MFCD00000158
Molecular Formula C5H11Br
Assay 97%
Health Hazard 3 GHS P Statement
P210-P280g-P273-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
UN Number UN1993
Beilstein 1730967
TSCA TSCA
Boiling Point 118° to 119°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 19°C (66°F)
Odor Characteristic
Quantity 5g
Health Hazard 1 Danger

Applications
3-Bromopentane is a glass forming liquid used in the preparation of 2-(1-ethyl-propoxy)-isoindole-1,3-dione. Further, it is employed in organic synthesis.

Solubility
Miscible with ethanol, ether, benzene and chloroform.

Notes
Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Danger

GHS P Statement
P210-P280g-P273-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.