Please login to your online account to display your discounted pricing

3-Bromopentane, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromopentane
Name Note Containing about 10% 2-Bromopentane
CAS 1809-10-5
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88% (typically), 10% (2-Bromopentane) typically
Assay Percent Range 90%
Molecular Formula C5H11Br
Linear Formula C2H5CH(Br)C2H5
Formula Weight 151.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389540250

  SDS
thermo fisher scientific
AC389540250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥88% (typically), 10% (2-Bromopentane) typically
Formula Weight 151.05
Quantity 25g
Color Clear pale yellow
Density 1.21g/mL
CAS Min % 88.0
CAS Max % 100.0
Chemical Name or Material 3-Bromopentane
Name Note Containing about 10% 2-Bromopentane
Assay Percent Range 90%
Molecular Formula C5H11Br
CAS 1809-10-5
Linear Formula C2H5CH(Br)C2H5
MDL Number MFCD00000158
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Packaging Glass bottle
Refractive Index 1.4420 to 1.4460 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.