missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Bromomethyl)thiophene, 97%

Manufacturer:  Alfa Aesar™ H6149703

 View more versions of this product

Catalog No. AAH6149703



Specifications

Specifications

3-(Bromomethyl)thiophene
1.6030
Brown
34846-44-1
MFCD03840497
3 bromomethyl thiophene
C1=CSC=C1CBr
177.059
177.06
97%
1.616g/mL
76°C (1mmHg)
1g
C5H5BrS
106459
KBWHYRUAHXHHFO-UHFFFAOYSA-N
3-(bromomethyl)thiophene
520703
Liquid

Chemical Identifiers

34846-44-1
177.059
KBWHYRUAHXHHFO-UHFFFAOYSA-N
520703
C1=CSC=C1CBr
C5H5BrS
MFCD03840497
3 bromomethyl thiophene
3-(bromomethyl)thiophene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents