missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Bromomethyl)thiophene, 97%

$109.95 - $109.95

Chemical Identifiers

CAS 34846-44-1
Formula Weight 177.06
MDL Number MFCD03840497
Beilstein 106459
Chemical Name or Material 3-(Bromomethyl)thiophene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6149703 SDS Alfa Aesar™
H6149703
1g Each for $109.95
AAH6149706 SDS Alfa Aesar™
H6149706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

34846-44-1
MFCD03840497
3-(Bromomethyl)thiophene
177.06
106459

Specifications

34846-44-1
1.616g/mL
Pale brown
1g
MFCD03840497
3-(Bromomethyl)thiophene
1.6030
76°C (1mmHg)
Liquid
177.06
106459
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C