Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Bromomethyl)benzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 3-(Bromomethyl)benzonitrile
CAS 28188-41-2
Molecular Formula C8H6BrN
Formula Weight 196.05
MDL Number MFCD00001809
UN Number UN2923
Beilstein 742356
Synonym alpha-Bromo-m-tolunitrile; 3-Cyanobenzyl bromide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1524406

  SDS
alfa aesar™
A1524406
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1524422

  SDS
alfa aesar™
A1524422
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.05
Chemical Name or Material 3-(Bromomethyl)benzonitrile
TSCA TSCA
Assay 98%
Molecular Formula C8H6BrN
MDL Number MFCD00001809
CAS 28188-41-2
UN Number UN2923
Melting Point 92° to 96°C
Beilstein 742356
Synonym alpha-Bromo-m-tolunitrile; 3-Cyanobenzyl bromide
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.