Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Bromomethyl)benzeneboronic acid, 95%

$25.86 - $25.86

Chemical Identifiers

Chemical Name or Material 3-(Bromomethyl)benzeneboronic acid
CAS 51323-43-4
Molecular Formula C7H8BBrO2
Formula Weight 214.85
MDL Number MFCD01632207
UN Number UN3261
Beilstein 3246618
Synonym 3-Boronobenzyl bromide; 3-(Bromomethyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2010206

  SDS
alfa aesar™
L2010206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2010214

  SDS
alfa aesar™
L2010214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2010203

  SDS
alfa aesar™
L2010203
1g Each for $25.86
Description & Specifications

Specifications

Chemical Name or Material 3-(Bromomethyl)benzeneboronic acid
CAS 51323-43-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 95%
Molecular Formula C7H8BBrO2
Formula Weight 214.85
MDL Number MFCD01632207
UN Number UN3261
Melting Point 214° to 216°C
Beilstein 3246618
Synonym 3-Boronobenzyl bromide; 3-(Bromomethyl)phenylboronic acid
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.