Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromomethyl-5-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Bromomethyl-5-fluorobenzeneboronic acid pinacol ester
Formula Weight 314.99
Synonym 3-Bromomethyl-5-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6215803

 
alfa aesar™
H6215803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62158MD

 
alfa aesar™
H62158MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.99
Assay 96%
Physical Form Solid
Quantity 1g
Chemical Name or Material 3-Bromomethyl-5-fluorobenzeneboronic acid pinacol ester
Synonym 3-Bromomethyl-5-fluorophenylboronic acid pinacol ester