missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Bromomethyl)-5-chlorobenzo[b]thiophene, TCI America™

$35.14 - $100.83

Chemical Identifiers

CAS 1198-51-2
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.561
MDL Number MFCD00173769
InChI Key FKQSFVITUNJLCY-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 3-bromomethyl-5-chlorobenzo b thiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, 3-bromomethyl-5-chlorobenzo b thiophene, pubchem13553
PubChem CID 2779857
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
SMILES C1=CC2=C(C=C1Cl)C(=CS2)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31471G View Documents TCI America
B31471G
1g Each for $35.14
 
B31475G View Documents TCI America
B31475G
5g Each for $100.83
 
Specifications

Chemical Identifiers

1198-51-2
261.561
FKQSFVITUNJLCY-UHFFFAOYSA-N
2779857
C1=CC2=C(C=C1Cl)C(=CS2)CBr
C9H6BrClS
MFCD00173769
3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 3-bromomethyl-5-chlorobenzo b thiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, 3-bromomethyl-5-chlorobenzo b thiophene, pubchem13553
3-(bromomethyl)-5-chloro-1-benzothiophene

Specifications

Undesignated
1g
C9H6BrClS
3261
FKQSFVITUNJLCY-UHFFFAOYSA-N
3-(bromomethyl)-5-chloro-1-benzothiophene
2779857
Crystal-Powder at 20°C
3-(Bromomethyl)-5-chlorobenzo[b]thiophene
131°C
1198-51-2
MFCD00173769
3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 3-bromomethyl-5-chlorobenzo b thiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, 3-bromomethyl-5-chlorobenzo b thiophene, pubchem13553
C1=CC2=C(C=C1Cl)C(=CS2)CBr
261.561
261.56
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator