missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromohexane, 97%

$67.54 - $67.54

Chemical Identifiers

CAS 3377-87-5
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
MDL Number MFCD00039181
InChI Key IOZOJWNUKLCDML-UHFFFAOYSA-N
Synonym hexane, 3-bromo, 3-hexyl bromide, acmc-1ct19, 3-bromohexane 5g, contains2-bromohexane stabilizedwithcopperchip
PubChem CID 18806
IUPAC Name 3-bromohexane
SMILES CCCC(CC)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0081806 View Documents Alfa Aesar™
L0081806
5g Each for $67.54
 
Description

Description

Applications
3-Bromohexane is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

3377-87-5
165.074
IOZOJWNUKLCDML-UHFFFAOYSA-N
18806
CCCC(CC)Br
C6H13Br
MFCD00039181
hexane, 3-bromo, 3-hexyl bromide, acmc-1ct19, 3-bromohexane 5g, contains2-bromohexane stabilizedwithcopperchip
3-bromohexane

Specifications

46°C (114°F)
144°C to 146°C
3377-87-5
MFCD00039181
1718967
IOZOJWNUKLCDML-UHFFFAOYSA-N
3-bromohexane
18806
97%
Pleasant
5g
C6H13Br
UN1993
hexane, 3-bromo, 3-hexyl bromide, acmc-1ct19, 3-bromohexane 5g, contains2-bromohexane stabilizedwithcopperchip
CCCC(CC)Br
165.074
165.08
3-Bromohexane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA