Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromoheptane, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromoheptane
CAS 1974-05-6
Molecular Formula C7H15Br
Formula Weight 179.11
MDL Number MFCD00039184
UN Number UN1993
Beilstein 1719072
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0442706

  SDS
alfa aesar™
L0442706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0442714

  SDS
alfa aesar™
L0442714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.11
Chemical Name or Material 3-Bromoheptane
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
CAS 1974-05-6
Beilstein 1719072
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Quantity 5g
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Boiling Point 61° to 62°C (18mmHg)
MDL Number MFCD00039184
Molecular Formula C7H15Br
Assay 97%
UN Number UN1993
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.