Learn More
3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2354314
Description
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
Specifications
3-Bromofuran | |
22037-28-1 | |
102°C to 104°C | |
C4H3BrO | |
MFCD00005347 | |
UN1993 | |
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
3-bromofuran | |
89164 | |
97% |
stabilized with 0.5% calcium carbonate | |
1.635 | |
3°C (37°F) | |
1.496 | |
25 g | |
105337 | |
Soluble in chloroform,ethyl acetate. Insoluble in water. | |
BrC1=COC=C1 | |
146.97 | |
146.98 |
Chemical Identifiers
22037-28-1 | |
146.97 | |
LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
89164 | |
BrC1=COC=C1 |
C4H3BrO | |
MFCD00005347 | |
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
3-bromofuran |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 244-744-6
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only