Please login to your online account to display your discounted pricing

3-Bromofuran, 97%, Maybridge


Quantity 25g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC02110EB

 View more versions of this product

Catalog No. CC02110EB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 146.97
Quantity 25g
Assay 95%
Density 1.635g/mL
MDL Number MFCD00005347
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%)
CAS 22037-28-1
CAS Max % 0.45
CAS 7440-50-8
CAS Max % 0.15
CAS Max % 0.55
CAS Min % 0.35
CAS Min % 94.5
CAS Max % 98.5
CAS 144-55-8
CAS Min % 0.05
CAS 110-00-9
CAS Min % 0.45
Packaging Amber glass bottle
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C4H3BrO
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%)
CAS 22037-28-1
CAS Min % 94.5
CAS Max % 98.5
CAS 7440-50-8
CAS Min % 0.05
CAS Max % 0.15
CAS 110-00-9
CAS Min % 0.45
CAS Max % 0.55
CAS 144-55-8
CAS Min % 0.35
CAS Max % 0.45
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3BrO
Formula Weight 146.97
MDL Number MFCD00005347
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents