missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromofuran, 97%, Maybridge™

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.971
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym .beta.-bromofuran, 3-bromo furan, 3-bromo-furan, 3-bromofuran, 3-furyl bromide, acmc-209fqz, beta-bromofuran, furan, 3-bromo, pubchem8830, zlchem 308
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES C1=COC=C1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC02110EA
SDS View Documents
Fisher Scientific Company
CC02110EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

22037-28-1
146.971
LXWLEQZDXOQZGW-UHFFFAOYSA-N
89164
C1=COC=C1Br
C4H3BrO
MFCD00005347
.beta.-bromofuran, 3-bromo furan, 3-bromo-furan, 3-bromofuran, 3-furyl bromide, acmc-209fqz, beta-bromofuran, furan, 3-bromo, pubchem8830, zlchem 308
3-bromofuran

Specifications

1.635g/mL
10g
94.5,0.05,0.45,0.35
95%
MFCD00005347
LXWLEQZDXOQZGW-UHFFFAOYSA-N
3-bromofuran
89164
95%
Amber glass bottle
22037-28-1,7440-50-8,110-00-9,144-55-8
98.5,0.15,0.55,0.45
C4H3BrO
.beta.-bromofuran, 3-bromo furan, 3-bromo-furan, 3-bromofuran, 3-furyl bromide, acmc-209fqz, beta-bromofuran, furan, 3-bromo, pubchem8830, zlchem 308
C1=COC=C1Br
146.971
146.97
3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%)
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : −20°C

SDS