Please login to your online account to display your discounted pricing

3-Bromofluorobenzene, 99%, ACROS Organics™


Quantity 500mL

Manufacturer: thermo fisher scientific  AC107005000

 View more versions of this product

Catalog No. AC107005000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 175
Physical Form Liquid
Quantity 500mL
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 214-023-0
RTECS Number DA1225000
CAS Max % 100.0
Synonym 1-Bromo-3-fluorobenzene
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Chemical Name or Material 3-Bromofluorobenzene
Assay Percent Range 99%
CAS 1073-06-9
Molecular Formula C6H4BrF
CAS Min % 98.5
Linear Formula BrC6H4F
Name Note 99%
Refractive Index 1.5260 to 1.5280 (20°C, 589nm)
MDL Number MFCD00000326
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Bromofluorobenzene
Name Note 99%
CAS 1073-06-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4BrF
Linear Formula BrC6H4F
Formula Weight 175
MDL Number MFCD00000326
Synonym 1-Bromo-3-fluorobenzene
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents