Please login to your online account to display your discounted pricing

3-Bromocytisine, Tocris™

Potent agonist of α4β4, α4β2 and α7 nACh receptors

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material 3-Bromocytisine
Assay Percent Range >98%
Molecular Formula C11H13BrN2O
Formula Weight 269.14
Synonym (1R,5S)-9-Bromo-1,2,3,4,5,6-hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

354910

 
r & d systems
354910
10mg Each for $257.85

354950

 
r & d systems
354950
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 269.14
Quantity 10mg
Chemical Name or Material 3-Bromocytisine
Assay Percent Range >98%
Molecular Formula C11H13BrN2O
Synonym (1R,5S)-9-Bromo-1,2,3,4,5,6-hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one
Recommended Storage Store at -20°C
CAS 207390-14-5
Safety and Handling

Store at -20°C