Learn More
3-Bromocyclohexene, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335120100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Bromocyclohexene | |
94.0 | |
White | |
64.0°C to 65.0°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5280 to 1.5320 | |
10 mL | |
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene | |
C1CC=CC(C1)Br | |
161.04 | |
161.04 | |
Solid |
1521-51-3 | |
100.0 | |
1.3800g/mL | |
54°C | |
94% min. (GC) | |
C6H9Br | |
MFCD00013775 | |
1.38 | |
AJKDUJRRWLQXHM-UHFFFAOYSA-N | |
3-bromocyclohexene | |
137057 | |
95% |
Chemical Identifiers
1521-51-3 | |
161.04 | |
AJKDUJRRWLQXHM-UHFFFAOYSA-N | |
137057 | |
C1CC=CC(C1)Br |
C6H9Br | |
MFCD00013775 | |
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene | |
3-bromocyclohexene |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
Recommended Storage : May darken on ageing
RUO – Research Use Only