missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromocumene 98.0+%, TCI America™
Supplier: TCI America B287525G
Specifications
3-Bromocumene | |
Yellow | |
C9H11Br | |
25 g | |
GBSGGFCCQZUXNB-UHFFFAOYSA-N | |
1-bromo-3-propan-2-ylbenzene | |
138499 | |
≥98.0% (GC) |
5433-01-2 | |
210°C | |
MFCD01318112 | |
1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene | |
CC(C)C1=CC(=CC=C1)Br | |
199.09 | |
199.09 | |
Liquid |
Chemical Identifiers
5433-01-2 | |
199.09 | |
GBSGGFCCQZUXNB-UHFFFAOYSA-N | |
138499 | |
CC(C)C1=CC(=CC=C1)Br |
C9H11Br | |
MFCD01318112 | |
1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene | |
1-bromo-3-propan-2-ylbenzene |
Safety and Handling
TSCA : No