missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromochromone, 97%

Manufacturer:  Alfa Aesar™ A1459101

 View more versions of this product

Catalog No. AAA1459101



Specifications

Specifications

3-Bromochromone
0.25g
C9H5BrO2
125684
IQIGYNPOESZBDJ-UHFFFAOYSA-N
3-bromochromen-4-one
521256
97%
94°C to 98°C
49619-82-1
MFCD00017337
3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, 3-bromochromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
C1=CC=C2C(=C1)C(=O)C(=CO2)Br
225.041
225.05

Chemical Identifiers

49619-82-1
225.041
IQIGYNPOESZBDJ-UHFFFAOYSA-N
521256
C1=CC=C2C(=C1)C(=O)C(=CO2)Br
C9H5BrO2
MFCD00017337
3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, 3-bromochromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
3-bromochromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents