missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromochlorobenzene, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 154201000

 View more versions of this product

Catalog No. AC154201000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Bromochlorobenzene
80°C
Glass bottle
1.63
Colorless to Yellow
100mL
108-37-2
100.0
C6H4BrCl
MFCD00000568
1-chloro-3-bromobenzene
JRGGUPZKKTVKOV-UHFFFAOYSA-N
1-bromo-3-chlorobenzene
7928
Liquid
1.6300g/mL
Authentic
1.5760 to 1.5780
195.0°C to 197.0°C
-21.5°C
99%
98.5
98.5% min. (GC)
BrC6H4Cl
05, 209
Solubility in water: insoluble
C1=CC(=CC(=C1)Br)Cl
191.45
191.45
99%

Chemical Identifiers

108-37-2
191.45
JRGGUPZKKTVKOV-UHFFFAOYSA-N
7928
C1=CC(=CC(=C1)Br)Cl
C6H4BrCl
MFCD00000568
1-chloro-3-bromobenzene
1-bromo-3-chlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 203-575-8

RTECSNumber : CY9005000

TSCA : TSCA

SDS Documents