missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-3-Bromocamphor, TCI America™

$50.42 - $385.02

Chemical Identifiers

CAS 10293-06-8
Molecular Formula C10H15BrO
Molecular Weight (g/mol) 231.133
MDL Number MFCD00003744
InChI Key NJQADTYRAYFBJN-MZVBAPTMSA-N
Synonym endo-3-bromo-d-camphor, +-camphor bromide, 3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
PubChem CID 44630109
IUPAC Name (1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2Br)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B056725G View Documents TCI America
B056725G
25g Each for $50.42
 
B0567500G View Documents TCI America
B0567500G
500g Each for $385.02
 
Specifications

Chemical Identifiers

10293-06-8
231.133
NJQADTYRAYFBJN-MZVBAPTMSA-N
44630109
CC1(C2CCC1(C(=O)C2Br)C)C
C10H15BrO
MFCD00003744
endo-3-bromo-d-camphor, +-camphor bromide, 3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
(1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one

Specifications

274°C
76°C
10293-06-8
MFCD00003744
NJQADTYRAYFBJN-MZVBAPTMSA-N
(1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
44630109
Crystal-Powder at 20°C
(+)-3-Bromocamphor
White
25g
C10H15BrO
endo-3-bromo-d-camphor, +-camphor bromide, 3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
CC1(C2CCC1(C(=O)C2Br)C)C
231.133
231.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0310

RTECSNumber : ED6750000

TSCA : Yes