Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-3-Bromocamphor, 98%

Chemical Identifiers

Chemical Name or Material (+)-3-Bromocamphor
CAS 10293-06-8
Formula Weight 231.13
MDL Number MFCD00003744
Beilstein 3197237
Synonym endo-(1R)-3-Bromocamphor; endo-(1R)-3-Bromo-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6601122

 
alfa aesar™
H6601122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6601136

 
alfa aesar™
H6601136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 75° to 77°C
Formula Weight 231.13
Chemical Name or Material (+)-3-Bromocamphor
Assay 98%
CAS 10293-06-8
Quantity 100g
Physical Form Crystalline
Sensitivity Light Sensitive
MDL Number MFCD00003744
Beilstein 3197237
Synonym endo-(1R)-3-Bromocamphor; endo-(1R)-3-Bromo-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Color White
RTECS Number ED6750000