missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt, TCI America™

$67.22 - $201.67

Chemical Identifiers

CAS 14575-84-9
Molecular Formula C10H18BrNO4S
Molecular Weight (g/mol) 328.221
MDL Number MFCD00167438
InChI Key GFBVBBRNPGPROZ-ZEMXJPTRSA-N
Synonym d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate
PubChem CID 71306887
IUPAC Name azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11255G View Documents TCI America
B11255G
5g Each for $67.22
 
B112525G View Documents TCI America
B112525G
25g Each for $201.67
 
Specifications

Chemical Identifiers

14575-84-9
328.221
GFBVBBRNPGPROZ-ZEMXJPTRSA-N
71306887
CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N
C10H18BrNO4S
MFCD00167438
d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate
azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid

Specifications

Undesignated
14575-84-9
MFCD00167438
GFBVBBRNPGPROZ-ZEMXJPTRSA-N
azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid
71306887
Crystal-Powder at 20°C
(+)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt
5g
C10H18BrNO4S
d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate
CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N
328.221
328.22
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No