missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobutyronitrile, TCI America™

$128.34 - $351.39

Chemical Identifiers

CAS 20965-20-2
Molecular Formula C4H6BrN
Molecular Weight (g/mol) 148.003
MDL Number MFCD00039488
InChI Key MBXSHBIQMDKTEW-UHFFFAOYSA-N
Synonym 2-Bromopropyl Cyanide
PubChem CID 549273
IUPAC Name 3-bromobutanenitrile
SMILES CC(CC#N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14525G View Documents TCI America
B14525G
5g Each for $128.34
 
B145225G View Documents TCI America
B145225G
25g Each for $351.39
 
Specifications

Chemical Identifiers

20965-20-2
148.003
MBXSHBIQMDKTEW-UHFFFAOYSA-N
549273
CC(CC#N)Br
C4H6BrN
MFCD00039488
2-Bromopropyl Cyanide
3-bromobutanenitrile

Specifications

72°C
5g
C4H6BrN
3276
MBXSHBIQMDKTEW-UHFFFAOYSA-N
3-bromobutanenitrile
549273
Clear Liquid at 20°C
3-Bromobutyronitrile
Yellow
20965-20-2
MFCD00039488
2-Bromopropyl Cyanide
CC(CC#N)Br
148.003
148.00
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No