missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobiphenyl, TCI America™

$74.86 - $675.30

Chemical Identifiers

CAS 2113-57-7
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.108
MDL Number MFCD00000082
InChI Key USYQKCQEVBFJRP-UHFFFAOYSA-N
Synonym 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
PubChem CID 16449
IUPAC Name 1-bromo-3-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20011G View Documents TCI America
B20011G
1g Each for $74.86
 
B20015G View Documents TCI America
B20015G
5g Each for $226.11
 
B200125G View Documents TCI America
B200125G
25g Each for $675.30
 
Specifications

Chemical Identifiers

2113-57-7
233.108
USYQKCQEVBFJRP-UHFFFAOYSA-N
16449
C1=CC=C(C=C1)C2=CC(=CC=C2)Br
C12H9Br
MFCD00000082
3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
1-bromo-3-phenylbenzene

Specifications

301°C
1g
C12H9Br
3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
C1=CC=C(C=C1)C2=CC(=CC=C2)Br
233.108
233.11
≥98.0% (GC)
Yellow
2113-57-7
MFCD00000082
USYQKCQEVBFJRP-UHFFFAOYSA-N
1-bromo-3-phenylbenzene
16449
Clear Liquid at 20°C
3-Bromobiphenyl
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0018

RTECSNumber : DV1750000

TSCA : Yes