Please login to your online account to display your discounted pricing

3-Bromobiphenyl, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  339100010

 View more versions of this product

Catalog No. AC339100010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Powder
Formula Weight 233.11
Quantity 1g
Color Light yellow to gray-beige
EINECS Number 218-304-9
TSCA TSCA
RTECS Number DV1750000
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Glass bottle
MDL Number MFCD00000082
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Bromobiphenyl
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H9Br
Linear Formula C6H5C6H4Br
CAS 2113-57-7
Infrared Spectrum Authentic
Refractive Index 1.6380 to 1.6420 (20°C, 589nm)

Specifications

Chemical Name or Material 3-Bromobiphenyl
Name Note 97%
CAS 2113-57-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C12H9Br
Linear Formula C6H5C6H4Br
Formula Weight 233.11
MDL Number MFCD00000082
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents