Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzylamine hydrochloride, 98%

$51.11 - $487.95

Chemical Identifiers

Chemical Name or Material 3-Bromobenzylamine hydrochloride
CAS 39959-54-1
Molecular Formula C7H8BrN·HCl
Formula Weight 222.52
MDL Number MFCD00012856
Beilstein 3913185
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1613206

  SDS
alfa aesar™
A1613206
5g Each for $51.11

AAA1613214

  SDS
alfa aesar™
A1613214
25g Each for $152.39

AAA1613222

  SDS
alfa aesar™
A1613222
100g Each for $487.95
Description & Specifications

Specifications

Formula Weight 222.52
Chemical Name or Material 3-Bromobenzylamine hydrochloride
Assay 98%
Molecular Formula C7H8BrN·HCl
MDL Number MFCD00012856
CAS 39959-54-1
Melting Point 218° to 221°C
Beilstein 3913185
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.