Learn More
3-Bromobenzylamine, 95%
CAS: 10269-01-9 | C7H8BrN | 186.05 g/mol
$66.49 - $66.49
Chemical Identifiers
| CAS | 10269-01-9 |
|---|---|
| Molecular Formula | C7H8BrN |
| Molecular Weight (g/mol) | 186.05 |
| MDL Number | MFCD01026119 |
| InChI Key | SUYJXERPRICYRX-UHFFFAOYSA-N |
| Synonym | 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 |
| PubChem CID | 457587 |
| IUPAC Name | (3-bromophenyl)methanamine |
| SMILES | C1=CC(=CC(=C1)Br)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC428960010
|
Thermo Scientific Chemicals
428960010 |
1 g | Glass bottle |
Each for $66.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10269-01-9 | |
| 186.05 | |
| SUYJXERPRICYRX-UHFFFAOYSA-N | |
| 457587 | |
| C1=CC(=CC(=C1)Br)CN |
| C7H8BrN | |
| MFCD01026119 | |
| 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 | |
| (3-bromophenyl)methanamine |
Specifications
| 10269-01-9 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 1.5850 to 1.5900 (20°C, 589nm) | |
| MFCD01026119 | |
| 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 | |
| C1=CC(=CC(=C1)Br)CN | |
| 186.05 | |
| 186.05 | |
| Liquid |
| 94.0 | |
| Colorless to Yellow | |
| 94% min. (GC) | |
| C7H8BrN | |
| BrC6H4CH2NH2 | |
| 1 g | |
| SUYJXERPRICYRX-UHFFFAOYSA-N | |
| (3-bromophenyl)methanamine | |
| 457587 | |
| 95% | |
| 3-Bromobenzylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only