missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzylamine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC428960010

 View more versions of this product

Catalog No. AC428960010


 

Specifications Description & Specifications

Specifications

3-Bromobenzylamine
Authentic
100.0
1.5850 to 1.5900 (20°C, 589nm)
1g
≥94%
C7H8BrN
186.05
10269-01-9
94.0
Glass bottle
Clear colorless to yellow
Liquid
95%
BrC6H4CH2NH2
MFCD01026119

Chemical Identifiers

10269-01-9
100.0
C7H8BrN
186.05
3-Bromobenzylamine
94.0
95%
BrC6H4CH2NH2
MFCD01026119
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger