missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzyl Alcohol 98.0+%, TCI America™

$61.38 - $182.49

Chemical Identifiers

CAS 15852-73-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004629
InChI Key FSWNRRSWFBXQCL-UHFFFAOYSA-N
Synonym 3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
PubChem CID 85141
IUPAC Name (3-bromophenyl)methanol
SMILES C1=CC(=CC(=C1)Br)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12875G
View Documents Promotion Details
TCI America
B12875G
5 g
Each for $61.38
 
B128725G
View Documents Promotion Details
TCI America
B128725G
25 g
Each for $182.49
 
Description

Description

Specifications

Chemical Identifiers

15852-73-0
187.036
FSWNRRSWFBXQCL-UHFFFAOYSA-N
85141
C1=CC(=CC(=C1)Br)CO
C7H7BrO
MFCD00004629
3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
(3-bromophenyl)methanol

Specifications

15852-73-0
165°C
MFCD00004629
3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
C1=CC(=CC(=C1)Br)CO
187.036
187.04
Liquid
Yellow
C7H7BrO
5 g
FSWNRRSWFBXQCL-UHFFFAOYSA-N
(3-bromophenyl)methanol
85141
≥98.0% (GC)
3-Bromobenzyl Alcohol
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'