Please login to your online account to display your discounted pricing

3-Bromobenzyl alcohol, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  182220050

 View more versions of this product

Catalog No. AC182220050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 187.04
Physical Form Liquid
Quantity 5g
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 239-975-4
Beilstein 06, 446
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5830 to 1.5850 (20°C, 589nm)
Molecular Formula C7H7BrO
Name Note 99%
MDL Number MFCD00004629
Assay Percent Range 99%
CAS 15852-73-0
CAS Min % 98.5
Linear Formula BrC6H4CH2OH
Chemical Name or Material 3-Bromobenzyl alcohol
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Bromobenzyl alcohol
Name Note 99%
CAS 15852-73-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7BrO
Linear Formula BrC6H4CH2OH
Formula Weight 187.04
MDL Number MFCD00004629
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents