Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzyl alcohol, 99%

$45.51 - $715.72

Chemical Identifiers

Chemical Name or Material 3-Bromobenzyl alcohol
CAS 15852-73-0
Molecular Formula C7H7BrO
Formula Weight 187.04
MDL Number MFCD00004629
Beilstein 2242512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1092914

  SDS
alfa aesar™
A1092914
25g Each for $188.35

AAA1092906

  SDS
alfa aesar™
A1092906
5g Each for $45.51

AAA1092922

  SDS
alfa aesar™
A1092922
100g Each for $715.72
Description & Specifications

Specifications

Formula Weight 187.04
Molecular Formula C7H7BrO
Quantity 25g
Assay 99%
Beilstein 2242512
MDL Number MFCD00004629
CAS 15852-73-0
Chemical Name or Material 3-Bromobenzyl alcohol
Boiling Point 165°C (16mmHg)
Flash Point >110°C (230°F)