missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzotrifluoride, 98+%

Manufacturer:  Alfa Aesar™ A1472436

 View more versions of this product

Catalog No. AAA1472436



Specifications

Specifications

3-Bromobenzotrifluoride
Aromatic
500g
C7H4BrF3
UN1993
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
C1=CC(=CC(=C1)Br)C(F)(F)F
225.008
225.01
59°C (138°F)
151°C to 152°C
401-78-5
MFCD00000380
1449557
NNMBNYHMJRJUBC-UHFFFAOYSA-N
1-bromo-3-(trifluoromethyl)benzene
9817
≥98%

Chemical Identifiers

401-78-5
225.008
NNMBNYHMJRJUBC-UHFFFAOYSA-N
9817
C1=CC(=CC(=C1)Br)C(F)(F)F
C7H4BrF3
MFCD00000380
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
1-bromo-3-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : XS7970000

TSCA : TSCA

SDS Documents